Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCACAACAAATGCCCACTGCTT[A/T]GGGTGACCTCAATTACACAAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF772 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF772 - zinc finger protein 772 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024596.2 | 6213 | UTR 3 | NP_001019767.1 | |||
NM_001144068.1 | 6213 | UTR 3 | NP_001137540.1 | |||
XM_005258943.4 | 6213 | UTR 3 | XP_005259000.1 | |||
XM_005258944.3 | 6213 | UTR 3 | XP_005259001.1 | |||
XM_011526980.2 | 6213 | UTR 3 | XP_011525282.1 | |||
XM_017026813.1 | 6213 | UTR 3 | XP_016882302.1 |