Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603363 | |||||||||||||||||||||||
Literature Links: |
CGGBP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CGGBP1 - CGG triplet repeat binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008390.1 | Intron | NP_001008391.1 | ||||
NM_001195308.1 | Intron | NP_001182237.1 | ||||
NM_003663.3 | Intron | NP_003654.3 | ||||
XM_017007355.1 | Intron | XP_016862844.1 |
ZNF654 - zinc finger protein 654 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018293.2 | Intron | NP_060763.2 | ||||
XM_011534340.2 | Intron | XP_011532642.2 | ||||
XM_017006789.1 | Intron | XP_016862278.1 | ||||
XM_017006790.1 | Intron | XP_016862279.1 | ||||
XM_017006791.1 | Intron | XP_016862280.1 | ||||
XM_017006792.1 | Intron | XP_016862281.1 | ||||
XM_017006793.1 | Intron | XP_016862282.1 | ||||
XM_017006794.1 | Intron | XP_016862283.1 | ||||
XM_017006795.1 | Intron | XP_016862284.1 |