Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTATGCATATCATTATCTGCCCC[C/T]ACCCAACCCACCAGCTCCCTGTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604520 | ||||||||||||||||||||
Literature Links: |
TNFSF14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNFSF14 - tumor necrosis factor superfamily member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003807.4 | 3280 | UTR 3 | NP_003798.2 | |||
NM_172014.3 | 3280 | UTR 3 | NP_742011.2 | |||
XM_017027417.1 | 3280 | UTR 3 | XP_016882906.1 | |||
XM_017027418.1 | 3280 | Intron | XP_016882907.1 |