Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAGACAACAGATGCTCAACAT[C/T]GATTAATATGTGTTAGGATTATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607384 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIMM9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TIMM9 - translocase of inner mitochondrial membrane 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304485.1 | Intron | NP_001291414.1 | ||||
NM_001304486.1 | Intron | NP_001291415.1 | ||||
NM_001304487.1 | Intron | NP_001291416.1 | ||||
NM_001304488.1 | Intron | NP_001291417.1 | ||||
NM_001304489.1 | Intron | NP_001291418.1 | ||||
NM_001304490.1 | Intron | NP_001291419.1 | ||||
NM_001304491.1 | Intron | NP_001291420.1 | ||||
NM_012460.3 | Intron | NP_036592.1 |
TOMM20L - translocase of outer mitochondrial membrane 20 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207377.2 | Intron | NP_997260.1 | ||||
XM_011536742.2 | Intron | XP_011535044.1 | ||||
XM_011536743.2 | Intron | XP_011535045.1 | ||||
XM_011536744.2 | Intron | XP_011535046.1 |
Set Membership: |
HapMap |