Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612137 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF146 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RNF146 - ring finger protein 146 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242844.1 | Intron | NP_001229773.1 | ||||
NM_001242845.1 | Intron | NP_001229774.1 | ||||
NM_001242846.1 | Intron | NP_001229775.1 | ||||
NM_001242847.1 | Intron | NP_001229776.1 | ||||
NM_001242848.1 | Intron | NP_001229777.1 | ||||
NM_001242849.1 | Intron | NP_001229778.1 | ||||
NM_001242850.1 | Intron | NP_001229779.1 | ||||
NM_001242851.1 | Intron | NP_001229780.1 | ||||
NM_001242852.1 | Intron | NP_001229781.1 | ||||
NM_030963.3 | Intron | NP_112225.2 | ||||
XM_006715571.3 | Intron | XP_006715634.1 | ||||
XM_011536161.2 | Intron | XP_011534463.1 | ||||
XM_011536162.2 | Intron | XP_011534464.1 | ||||
XM_011536163.2 | Intron | XP_011534465.1 | ||||
XM_011536164.2 | Intron | XP_011534466.1 | ||||
XM_017011336.1 | Intron | XP_016866825.1 | ||||
XM_017011337.1 | Intron | XP_016866826.1 | ||||
XM_017011338.1 | Intron | XP_016866827.1 | ||||
XM_017011339.1 | Intron | XP_016866828.1 | ||||
XM_017011340.1 | Intron | XP_016866829.1 | ||||
XM_017011341.1 | Intron | XP_016866830.1 | ||||
XM_017011342.1 | Intron | XP_016866831.1 | ||||
XM_017011343.1 | Intron | XP_016866832.1 |