Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605532 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP95 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP95 - centrosomal protein 95 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMURF2 - SMAD specific E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022739.3 | 2892 | UTR 3 | NP_073576.1 | |||
XM_005257585.3 | 2892 | UTR 3 | XP_005257642.1 | |||
XM_011525128.1 | 2892 | UTR 3 | XP_011523430.1 |
Set Membership: |
HapMap |