Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGGAGTTTTGGAATAGGACATC[C/T]CACATCCGTGGACACACATGTAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615894 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00909 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LINC00909 - long intergenic non-protein coding RNA 909 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF407 - zinc finger protein 407 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146189.1 | Intron | NP_001139661.1 | ||||
NM_001146190.1 | Intron | NP_001139662.1 | ||||
NM_017757.2 | Intron | NP_060227.2 | ||||
XM_005266726.4 | Intron | XP_005266783.1 | ||||
XM_006722500.3 | Intron | XP_006722563.1 | ||||
XM_011526068.2 | Intron | XP_011524370.1 | ||||
XM_011526069.2 | Intron | XP_011524371.1 | ||||
XM_011526070.1 | Intron | XP_011524372.1 | ||||
XM_017025838.1 | Intron | XP_016881327.1 |
Set Membership: |
HapMap |