Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608072 MIM: 187680 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NHLRC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NHLRC1 - NHL repeat containing E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPMT - thiopurine S-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000367.3 | 3304 | UTR 3 | NP_000358.1 | |||
XM_011514839.2 | 3304 | UTR 3 | XP_011513141.1 | |||
XM_017011241.1 | 3304 | UTR 3 | XP_016866730.1 |
Set Membership: |
HapMap |