Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601965 MIM: 605263 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
S1PR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
S1PR3 - sphingosine-1-phosphate receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHC3 - SHC adaptor protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016848.5 | 7763 | UTR 3 | NP_058544.3 | |||
XM_011518785.2 | 7763 | UTR 3 | XP_011517087.1 | |||
XM_011518786.2 | 7763 | UTR 3 | XP_011517088.1 | |||
XM_017014809.1 | 7763 | Intron | XP_016870298.1 | |||
XM_017014810.1 | 7763 | Intron | XP_016870299.1 |
Set Membership: |
HapMap |