Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCACTTTCTGTCTTTCTTTTAT[G/T]TAACATCTCTGCAGCACTGACATTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601987 MIM: 615775 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHKB-CPT1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CHKB-CPT1B - CHKB-CPT1B readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPT1B - carnitine palmitoyltransferase 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLHDC7B - kelch domain containing 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYCE3 - synaptonemal complex central element protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |