Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCACAGAGAAAGGAGGTTGGC[C/T]GAAGACTGATTAGGTTTACATGAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602287 MIM: 612755 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERP29 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ERP29 - endoplasmic reticulum protein 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3657 - microRNA 3657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAA25 - N(alpha)-acetyltransferase 25, NatB auxiliary subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024953.3 | Intron | NP_079229.2 | ||||
XM_005253938.1 | Intron | XP_005253995.1 | ||||
XM_006719606.2 | Intron | XP_006719669.1 | ||||
XM_017019977.1 | Intron | XP_016875466.1 |
Set Membership: |
HapMap |