Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606193 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC13A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC13A1 - solute carrier family 13 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324400.1 | 3528 | UTR 3 | NP_001311329.1 | |||
NM_022444.3 | 3528 | UTR 3 | NP_071889.2 | |||
XM_011516515.2 | 3528 | UTR 3 | XP_011514817.1 | |||
XM_011516516.2 | 3528 | UTR 3 | XP_011514818.1 | |||
XM_011516517.2 | 3528 | UTR 3 | XP_011514819.1 | |||
XM_011516518.2 | 3528 | Intron | XP_011514820.1 | |||
XM_011516519.2 | 3528 | Intron | XP_011514821.1 | |||
XM_017012554.1 | 3528 | UTR 3 | XP_016868043.1 | |||
XM_017012555.1 | 3528 | Intron | XP_016868044.1 |
Set Membership: |
HapMap DME Validated Inventoried |