Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300350 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRDL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHRDL1 - chordin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143981.1 | Intron | NP_001137453.1 | ||||
NM_001143982.1 | Intron | NP_001137454.1 | ||||
NM_001143983.2 | Intron | NP_001137455.2 | ||||
NM_145234.3 | Intron | NP_660277.2 | ||||
XM_005262221.1 | Intron | XP_005262278.1 | ||||
XM_005262222.3 | Intron | XP_005262279.1 | ||||
XM_005262223.1 | Intron | XP_005262280.1 | ||||
XM_005262224.1 | Intron | XP_005262281.1 | ||||
XM_017029959.1 | Intron | XP_016885448.1 |