Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606751 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPRSS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMPRSS5 - transmembrane protease, serine 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288749.1 | 5910 | Missense Mutation | CTC,TTC | L,F 256 | NP_001275678.1 | |
NM_001288750.1 | 5910 | Missense Mutation | CTC,TTC | L,F 325 | NP_001275679.1 | |
NM_001288751.1 | 5910 | Missense Mutation | CTC,TTC | L,F 360 | NP_001275680.1 | |
NM_001288752.1 | 5910 | Missense Mutation | CTC,TTC | L,F 300 | NP_001275681.1 | |
NM_030770.3 | 5910 | Missense Mutation | CTC,TTC | L,F 369 | NP_110397.2 | |
XM_017018366.1 | 5910 | Missense Mutation | CTC,TTC | L,F 139 | XP_016873855.1 | |
XM_017018367.1 | 5910 | Missense Mutation | CTC,TTC | L,F 139 | XP_016873856.1 |
Set Membership: |
HapMap |