Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 604105 | ||||||||||||||||||||||||||
Literature Links: |
SYCP2 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
SYCP2 - synaptonemal complex protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014258.3 | Intron | NP_055073.2 | ||||
XM_011528487.2 | Intron | XP_011526789.1 | ||||
XM_011528488.2 | Intron | XP_011526790.1 | ||||
XM_011528489.2 | Intron | XP_011526791.1 | ||||
XM_011528490.2 | Intron | XP_011526792.1 | ||||
XM_011528493.2 | Intron | XP_011526795.1 | ||||
XM_017027586.1 | Intron | XP_016883075.1 | ||||
XM_017027587.1 | Intron | XP_016883076.1 | ||||
XM_017027588.1 | Intron | XP_016883077.1 | ||||
XM_017027589.1 | Intron | XP_016883078.1 | ||||
XM_017027590.1 | Intron | XP_016883079.1 | ||||
XM_017027591.1 | Intron | XP_016883080.1 | ||||
XM_017027592.1 | Intron | XP_016883081.1 |
Set Membership: |
HapMap |