Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH26 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDH26 - cadherin 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021810.4 | 923 | Intron | NP_068582.2 | |||
NM_177980.2 | 923 | UTR 5 | NP_817089.1 | |||
XM_011528969.2 | 923 | UTR 5 | XP_011527271.1 | |||
XM_011528970.2 | 923 | Intron | XP_011527272.1 | |||
XM_017027994.1 | 923 | UTR 5 | XP_016883483.1 | |||
XM_017027995.1 | 923 | UTR 5 | XP_016883484.1 | |||
XM_017027996.1 | 923 | UTR 5 | XP_016883485.1 | |||
XM_017027997.1 | 923 | UTR 5 | XP_016883486.1 | |||
XM_017027998.1 | 923 | UTR 5 | XP_016883487.1 | |||
XM_017027999.1 | 923 | UTR 5 | XP_016883488.1 | |||
XM_017028000.1 | 923 | UTR 5 | XP_016883489.1 | |||
XM_017028001.1 | 923 | UTR 5 | XP_016883490.1 | |||
XM_017028002.1 | 923 | UTR 5 | XP_016883491.1 |
Set Membership: |
HapMap |