Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTAGAGAAGTGGCATTGGTTTT[C/T]ATTTTGAAGATCTCTTTAATGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604860 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927322 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101927322 - uncharacterized LOC101927322 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289967.1 | Intron | NP_001276896.1 |
LOC102724745 - uncharacterized LOC102724745 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MALT1 - MALT1 paracaspase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006785.3 | Intron | NP_006776.1 | ||||
NM_173844.2 | Intron | NP_776216.1 | ||||
XM_011525794.1 | Intron | XP_011524096.1 |
Set Membership: |
HapMap |