Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAGTGTTGAATTTAATTTCAGGA[C/T]TGCTTTATTACAGTTTTCTTTCCTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605787 MIM: 616194 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRA2 - ankyrin repeat family A member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UTP15 - UTP15, small subunit processome component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284430.1 | Intron | NP_001271359.1 | ||||
NM_001284431.1 | Intron | NP_001271360.1 | ||||
NM_032175.3 | Intron | NP_115551.2 | ||||
XM_011543680.2 | Intron | XP_011541982.1 |
Set Membership: |
HapMap |