Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCTGGGTGCACCACCCCTGGGC[C/T]TTCGGCGACGCCGGCTGCCGCGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 162651 | ||||||||||||||||||||
Literature Links: |
LOC107987285 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107987285 - uncharacterized LOC107987285 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NTSR1 - neurotensin receptor 1 (high affinity) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002531.2 | 776 | Silent Mutation | GCC,GCT | A,A 135 | NP_002522.2 | |
XM_011528827.2 | 776 | Silent Mutation | GCC,GCT | A,A 135 | XP_011527129.1 |