Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCT - dopachrome tautomerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129889.2 | Intron | NP_001123361.1 | ||||
NM_001322182.1 | Intron | NP_001309111.1 | ||||
NM_001322183.1 | Intron | NP_001309112.1 | ||||
NM_001322184.1 | Intron | NP_001309113.1 | ||||
NM_001322185.1 | Intron | NP_001309114.1 | ||||
NM_001322186.1 | Intron | NP_001309115.1 | ||||
NM_001922.4 | Intron | NP_001913.2 | ||||
XM_011521049.1 | Intron | XP_011519351.1 | ||||
XM_017020401.1 | Intron | XP_016875890.1 |