Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCATCATTTGTGCCTGTTTATG[C/T]TTATATGCACAAATGCACTAATTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612139 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PREX2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PREX2 - phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024870.3 | Intron | NP_079146.2 | ||||
NM_025170.5 | Intron | NP_079446.3 | ||||
XM_011517612.2 | Intron | XP_011515914.1 |
Set Membership: |
HapMap |