Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616565 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD30B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD30B - ankyrin repeat domain 30B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145029.1 | Intron | NP_001138501.1 | ||||
XM_006722325.3 | Intron | XP_006722388.1 | ||||
XM_011525662.2 | Intron | XP_011523964.1 | ||||
XM_011525664.2 | Intron | XP_011523966.1 | ||||
XM_011525665.2 | Intron | XP_011523967.1 | ||||
XM_011525666.2 | Intron | XP_011523968.1 |