Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTCACCTGTCCCAATTGCGCA[C/G]CCTCGTGTCCACCCACAAACCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608661 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PODN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PODN - podocan | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153703.4 | Intron | NP_714914.2 | ||||
XM_005270457.3 | Intron | XP_005270514.1 | ||||
XM_011540677.2 | Intron | XP_011538979.1 | ||||
XM_017000294.1 | Intron | XP_016855783.1 |
Set Membership: |
HapMap |