Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCACTGGGGAAAAGATGATTTTA[C/T]AGCTTACAACCACATGGCATGTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601411 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGCD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SGCD - sarcoglycan delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000337.5 | Intron | NP_000328.2 | ||||
NM_001128209.1 | Intron | NP_001121681.1 | ||||
NM_172244.2 | Intron | NP_758447.1 | ||||
XM_005265966.4 | Intron | XP_005266023.1 | ||||
XM_005265967.2 | Intron | XP_005266024.1 | ||||
XM_011534621.2 | Intron | XP_011532923.1 | ||||
XM_017009723.1 | Intron | XP_016865212.1 | ||||
XM_017009724.1 | Intron | XP_016865213.1 |
Set Membership: |
HapMap |