Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605795 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEX15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TEX15 - testis expressed 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031271.3 | Intron | NP_112561.2 | ||||
XM_005273575.3 | Intron | XP_005273632.1 | ||||
XM_005273576.3 | Intron | XP_005273633.1 | ||||
XM_006716369.3 | Intron | XP_006716432.1 | ||||
XM_011544588.2 | Intron | XP_011542890.1 | ||||
XM_011544589.2 | Intron | XP_011542891.1 | ||||
XM_011544590.2 | Intron | XP_011542892.1 | ||||
XM_011544591.2 | Intron | XP_011542893.1 | ||||
XM_011544592.2 | Intron | XP_011542894.1 | ||||
XM_011544593.2 | Intron | XP_011542895.1 | ||||
XM_011544594.2 | Intron | XP_011542896.1 |