Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615661 MIM: 613295 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCD2L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDCD2L - programmed cell death 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBA2 - ubiquitin like modifier activating enzyme 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005499.2 | Intron | NP_005490.1 | ||||
XM_005258404.3 | Intron | XP_005258461.2 | ||||
XM_006722962.2 | Intron | XP_006723025.1 | ||||
XM_011526304.2 | Intron | XP_011524606.1 | ||||
XM_017026134.1 | Intron | XP_016881623.1 |
Set Membership: |
HapMap |