Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604545 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KPNA5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KPNA5 - karyopherin subunit alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002269.2 | 29 | Intron | NP_002260.2 | |||
XM_011535805.1 | 29 | UTR 5 | XP_011534107.1 | |||
XM_017010838.1 | 29 | UTR 5 | XP_016866327.1 | |||
XM_017010839.1 | 29 | Intron | XP_016866328.1 | |||
XM_017010840.1 | 29 | Intron | XP_016866329.1 | |||
XM_017010841.1 | 29 | Intron | XP_016866330.1 | |||
XM_017010842.1 | 29 | Intron | XP_016866331.1 | |||
XM_017010843.1 | 29 | Intron | XP_016866332.1 |
Set Membership: |
HapMap |