Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGTTATTTTCAAGCTTTGCTAAC[A/G]GTAGGTGATAAAATTGACAAGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606808 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929073 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929073 - uncharacterized LOC101929073 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYO3A - myosin IIIA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017433.4 | 187 | UTR 5 | NP_059129.3 | |||
XM_011519498.2 | 187 | UTR 5 | XP_011517800.1 | |||
XM_011519499.1 | 187 | UTR 5 | XP_011517801.1 | |||
XM_011519500.2 | 187 | UTR 5 | XP_011517802.1 | |||
XM_011519502.1 | 187 | UTR 5 | XP_011517804.1 | |||
XM_011519503.1 | 187 | UTR 5 | XP_011517805.1 | |||
XM_011519504.1 | 187 | UTR 5 | XP_011517806.1 | |||
XM_011519505.1 | 187 | UTR 5 | XP_011517807.1 | |||
XM_011519506.2 | 187 | UTR 5 | XP_011517808.1 | |||
XM_011519508.2 | 187 | UTR 5 | XP_011517810.1 | |||
XM_011519510.1 | 187 | UTR 5 | XP_011517812.1 | |||
XM_011519511.2 | 187 | UTR 5 | XP_011517813.1 | |||
XM_011519512.1 | 187 | Intron | XP_011517814.1 | |||
XM_011519513.2 | 187 | Intron | XP_011517815.1 |