Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAAAAGCTGAGCTAAAGGAATGA[A/G]ACACAAAAGAGTACACATACTGTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613056 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf55-LUC7L2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C7orf55-LUC7L2 - C7orf55-LUC7L2 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLRG2 - killer cell lectin like receptor G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198508.2 | Intron | NP_940910.1 | ||||
XM_005250311.3 | Intron | XP_005250368.1 | ||||
XM_011516140.2 | Intron | XP_011514442.1 | ||||
XM_011516141.2 | Intron | XP_011514443.1 |
LOC100129148 - uncharacterized LOC100129148 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LUC7L2 - LUC7 like 2, pre-mRNA splicing factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |