Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTTGTAACGCAGAACCTGTGAA[C/G]AAGTTCTTTCATTTCATAGTATTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604517 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPARGC1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPARGC1A - PPARG coactivator 1 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013261.3 | 4714 | UTR 3 | NP_037393.1 | |||
XM_005248131.4 | 4714 | UTR 3 | XP_005248188.1 | |||
XM_005248132.1 | 4714 | UTR 3 | XP_005248189.1 | |||
XM_005248134.4 | 4714 | UTR 3 | XP_005248191.1 | |||
XM_011513765.2 | 4714 | UTR 3 | XP_011512067.1 | |||
XM_011513766.1 | 4714 | UTR 3 | XP_011512068.1 | |||
XM_011513767.2 | 4714 | UTR 3 | XP_011512069.1 | |||
XM_011513768.1 | 4714 | UTR 3 | XP_011512070.1 | |||
XM_011513769.2 | 4714 | Intron | XP_011512071.1 | |||
XM_011513770.2 | 4714 | UTR 3 | XP_011512072.1 | |||
XM_011513771.1 | 4714 | UTR 3 | XP_011512073.1 | |||
XM_017007664.1 | 4714 | UTR 3 | XP_016863153.1 |
Set Membership: |
HapMap |