Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCTGTCCCCTTTACGCCAGGGT[C/T]TCTCCACCTGGGCACGGTTGGTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612939 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HSPBP1 - HSPA (Hsp70) binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130106.1 | Intron | NP_001123578.1 | ||||
NM_001297600.1 | Intron | NP_001284529.1 | ||||
NM_012267.4 | Intron | NP_036399.3 | ||||
XM_005258700.2 | Intron | XP_005258757.1 | ||||
XM_005258701.2 | Intron | XP_005258758.1 | ||||
XM_011526691.1 | Intron | XP_011524993.1 | ||||
XM_017026543.1 | Intron | XP_016882032.1 | ||||
XM_017026544.1 | Intron | XP_016882033.1 | ||||
XM_017026545.1 | Intron | XP_016882034.1 |