Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607167 MIM: 606409 | ||||||||||||||||||||
Literature Links: |
DYNLRB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DYNLRB1 - dynein light chain roadblock-type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281727.1 | Intron | NP_001268656.1 | ||||
NM_001281728.1 | Intron | NP_001268657.1 | ||||
NM_001281729.1 | Intron | NP_001268658.1 | ||||
NM_001319157.1 | Intron | NP_001306086.1 | ||||
NM_014183.3 | Intron | NP_054902.1 | ||||
NM_177953.2 | Intron | NP_808852.1 |
ITCH - itchy E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |