Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102910 MIM: 605682 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP5B - ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001686.3 | Intron | NP_001677.2 |
BAZ2A - bromodomain adjacent to zinc finger domain 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD59A - small nucleolar RNA, C/D box 59A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD59B - small nucleolar RNA, C/D box 59B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |