Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300820 MIM: 300502 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP3K15 - mitogen-activated protein kinase kinase kinase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001671.3 | 2046 | UTR 3 | NP_001001671.3 | |||
XM_011545507.2 | 2046 | UTR 3 | XP_011543809.2 | |||
XM_011545508.2 | 2046 | UTR 3 | XP_011543810.2 | |||
XM_011545510.2 | 2046 | UTR 3 | XP_011543812.1 | |||
XM_011545511.2 | 2046 | UTR 3 | XP_011543813.1 | |||
XM_017029511.1 | 2046 | Intron | XP_016885000.1 |
PDHA1 - pyruvate dehydrogenase (lipoamide) alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000284.3 | 2046 | UTR 3 | NP_000275.1 | |||
NM_001173454.1 | 2046 | UTR 3 | NP_001166925.1 | |||
NM_001173455.1 | 2046 | UTR 3 | NP_001166926.1 | |||
NM_001173456.1 | 2046 | UTR 3 | NP_001166927.1 | |||
XM_017029574.1 | 2046 | Intron | XP_016885063.1 |
Set Membership: |
HapMap |