Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109770 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEACAM1 - carcinoembryonic antigen related cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024912.2 | Intron | NP_001020083.1 | ||||
NM_001184813.1 | Intron | NP_001171742.1 | ||||
NM_001184815.1 | Intron | NP_001171744.1 | ||||
NM_001184816.1 | Intron | NP_001171745.1 | ||||
NM_001205344.1 | Intron | NP_001192273.1 | ||||
NM_001712.4 | Intron | NP_001703.2 | ||||
XM_011527206.1 | Intron | XP_011525508.1 |
LIPE-AS1 - LIPE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |