Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604100 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRM4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GRM4 - glutamate metabotropic receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000841.3 | Intron | NP_000832.1 | ||||
NM_001256809.2 | Intron | NP_001243738.1 | ||||
NM_001256811.2 | Intron | NP_001243740.1 | ||||
NM_001256812.2 | Intron | NP_001243741.1 | ||||
NM_001256813.2 | Intron | NP_001243742.1 | ||||
NM_001282847.1 | Intron | NP_001269776.1 | ||||
XM_017010790.1 | Intron | XP_016866279.1 | ||||
XM_017010791.1 | Intron | XP_016866280.1 | ||||
XM_017010792.1 | Intron | XP_016866281.1 | ||||
XM_017010793.1 | Intron | XP_016866282.1 |
Set Membership: |
HapMap |