Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612377 MIM: 603090 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COMMD6 - COMM domain containing 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UCHL3 - ubiquitin C-terminal hydrolase L3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270952.1 | Intron | NP_001257881.1 | ||||
NM_006002.4 | Intron | NP_005993.1 | ||||
XM_011535212.1 | Intron | XP_011533514.1 | ||||
XM_011535213.2 | Intron | XP_011533515.1 | ||||
XM_011535214.2 | Intron | XP_011533516.1 | ||||
XM_017020725.1 | Intron | XP_016876214.1 | ||||
XM_017020726.1 | Intron | XP_016876215.1 | ||||
XM_017020727.1 | Intron | XP_016876216.1 |
Set Membership: |
HapMap |