Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607144 MIM: 607171 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALG12 - ALG12, alpha-1,6-mannosyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRELD2 - cysteine rich with EGF like domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135101.2 | 413 | Silent Mutation | AAC,AAT | N,N 91 | NP_001128573.1 | |
NM_001284317.1 | 413 | Silent Mutation | AAC,AAT | N,N 91 | NP_001271246.1 | |
NM_001284318.1 | 413 | Silent Mutation | AAC,AAT | N,N 91 | NP_001271247.1 | |
NM_024324.4 | 413 | Silent Mutation | AAC,AAT | N,N 91 | NP_077300.3 | |
XM_005261737.2 | 413 | Silent Mutation | AAC,AAT | N,N 91 | XP_005261794.1 | |
XM_005261738.4 | 413 | Silent Mutation | AAC,AAT | N,N 91 | XP_005261795.1 | |
XM_011530372.2 | 413 | Silent Mutation | AAC,AAT | N,N 91 | XP_011528674.1 | |
XM_017028938.1 | 413 | UTR 5 | XP_016884427.1 |
Set Membership: |
HapMap |