Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAGCTCTAAAGTCAGGTAATG[G/T]TTTTCATGTGCTTCTCTTGAGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
59 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146880 MIM: 604305 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-DQA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
SAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
EAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
||||||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR - Not Available | ||||||||
HLA-DQA1 - major histocompatibility complex, class II, DQ alpha 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HLA-DQB1 - major histocompatibility complex, class II, DQ beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243961.1 | 1167 | UTR 3 | NP_001230890.1 | |||
NM_001243962.1 | 1167 | UTR 3 | NP_001230891.1 | |||
NM_002123.4 | 1167 | UTR 3 | NP_002114.3 |
HLA-DQB1-AS1 - HLA-DQB1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |