Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACACAAGAAAGTCTTATGTAGT[A/G]AATTTTTGTCCTGAAATAAAATAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LILRB1 - leukocyte immunoglobulin like receptor B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081637.2 | Intron | NP_001075106.2 | ||||
NM_001081638.3 | Intron | NP_001075107.2 | ||||
NM_001081639.3 | Intron | NP_001075108.2 | ||||
NM_001278398.2 | Intron | NP_001265327.2 | ||||
NM_001278399.2 | Intron | NP_001265328.2 | ||||
NM_006669.6 | Intron | NP_006660.4 | ||||
XM_011526331.2 | Intron | XP_011524633.1 | ||||
XM_011526332.2 | Intron | XP_011524634.1 | ||||
XM_011526335.2 | Intron | XP_011524637.1 | ||||
XM_011526336.2 | Intron | XP_011524638.1 | ||||
XM_011526338.2 | Intron | XP_011524640.1 | ||||
XM_017026182.1 | Intron | XP_016881671.1 | ||||
XM_017026183.1 | Intron | XP_016881672.1 | ||||
XM_017026184.1 | Intron | XP_016881673.1 | ||||
XM_017026185.1 | Intron | XP_016881674.1 | ||||
XM_017026186.1 | Intron | XP_016881675.1 | ||||
XM_017026187.1 | Intron | XP_016881676.1 | ||||
XM_017026188.1 | Intron | XP_016881677.1 | ||||
XM_017026189.1 | Intron | XP_016881678.1 | ||||
XM_017026190.1 | Intron | XP_016881679.1 | ||||
XM_017026191.1 | Intron | XP_016881680.1 | ||||
XM_017026192.1 | Intron | XP_016881681.1 |