Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K8 - mitogen-activated protein kinase kinase kinase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244134.1 | 500 | Intron | NP_001231063.1 | |||
NM_001320961.1 | 500 | Intron | NP_001307890.1 | |||
NM_005204.3 | 500 | Intron | NP_005195.2 | |||
XM_017015707.1 | 500 | Intron | XP_016871196.1 | |||
XM_017015708.1 | 500 | Intron | XP_016871197.1 | |||
XM_017015709.1 | 500 | UTR 5 | XP_016871198.1 | |||
XM_017015710.1 | 500 | Intron | XP_016871199.1 | |||
XM_017015711.1 | 500 | Intron | XP_016871200.1 | |||
XM_017015712.1 | 500 | Intron | XP_016871201.1 | |||
XM_017015713.1 | 500 | Intron | XP_016871202.1 | |||
XM_017015714.1 | 500 | Intron | XP_016871203.1 |