Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCTAGAGGCAGAGCCTATTCTA[C/T]CCTGGGCATCGCTGACCCTCTGCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610823 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARMC6 - armadillo repeat containing 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A42 - solute carrier family 25 member 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321544.1 | Intron | NP_001308473.1 | ||||
NM_178526.4 | Intron | NP_848621.2 | ||||
XM_005259861.3 | Intron | XP_005259918.1 | ||||
XM_011527937.2 | Intron | XP_011526239.1 | ||||
XM_017026670.1 | Intron | XP_016882159.1 |
Set Membership: |
HapMap |