Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCAGCCTGGGCAACATAGCAAGA[C/T]GTCGTCTCCATAAAAATATTAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609063 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TXN2 - thioredoxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012473.3 | Intron | NP_036605.2 | ||||
XM_005261508.1 | Intron | XP_005261565.1 | ||||
XM_006724226.1 | Intron | XP_006724289.1 | ||||
XM_011530111.2 | Intron | XP_011528413.1 |
Set Membership: |
HapMap |