Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACGGGAGTCGCCGTGAGCACCA[A/G]CATCTCCAGTCCCTTCAGGCCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605140 MIM: 609108 | ||||||||||||||||||||
Literature Links: |
CCT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCT7 - chaperonin containing TCP1 subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO41 - F-box protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080410.2 | 3382 | Silent Mutation | CTG,TTG | L,L 800 | NP_001073879.2 | |
XM_005264158.3 | 3382 | Silent Mutation | CTG,TTG | L,L 406 | XP_005264215.1 | |
XM_006711950.2 | 3382 | Silent Mutation | CTG,TTG | L,L 800 | XP_006712013.1 | |
XM_011532562.1 | 3382 | Silent Mutation | CTG,TTG | L,L 800 | XP_011530864.1 | |
XM_011532563.2 | 3382 | Silent Mutation | CTG,TTG | L,L 552 | XP_011530865.1 | |
XM_017003421.1 | 3382 | Silent Mutation | CTG,TTG | L,L 800 | XP_016858910.1 | |
XM_017003422.1 | 3382 | Silent Mutation | CTG,TTG | L,L 800 | XP_016858911.1 | |
XM_017003423.1 | 3382 | Silent Mutation | CTG,TTG | L,L 800 | XP_016858912.1 |