Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MX2 - MX dynamin like GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002463.1 | Intron | NP_002454.1 | ||||
XM_005260983.4 | Intron | XP_005261040.1 | ||||
XM_005260984.1 | Intron | XP_005261041.1 | ||||
XM_011529571.1 | Intron | XP_011527873.1 | ||||
XM_011529572.1 | Intron | XP_011527874.1 | ||||
XM_011529573.1 | Intron | XP_011527875.1 | ||||
XM_011529574.2 | Intron | XP_011527876.1 |