Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATGTGGAGGAAGGTGAACTGGA[G/T]TGCCTGAAGGGTGAGATACTGCAGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606833 | |||||||||||||||||||||||
Literature Links: |
KMT2C PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LOC731075 - uncharacterized LOC731075 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YBX1P4 - Y-box binding protein 1 pseudogene 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |