Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGTCCAGAGACTTAACCCACA[A/G]CACTTGTCGACACTGTGGAACACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609226 | ||||||||||||||||||||
Literature Links: |
WDR45B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR45B - WD repeat domain 45B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019613.3 | Intron | NP_062559.2 | ||||
XM_005256377.4 | Intron | XP_005256434.1 | ||||
XM_017024862.1 | Intron | XP_016880351.1 | ||||
XM_017024863.1 | Intron | XP_016880352.1 |