Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616236 MIM: 611865 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHADL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHADL - chondroadherin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EP300-AS1 - EP300 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
L3MBTL2 - L3MBTL2 polycomb repressive complex 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031488.4 | Intron | NP_113676.2 | ||||
XM_011530420.2 | Intron | XP_011528722.1 | ||||
XM_011530421.2 | Intron | XP_011528723.1 | ||||
XM_017028976.1 | Intron | XP_016884465.1 | ||||
XM_017028977.1 | Intron | XP_016884466.1 | ||||
XM_017028978.1 | Intron | XP_016884467.1 | ||||
XM_017028979.1 | Intron | XP_016884468.1 | ||||
XM_017028980.1 | Intron | XP_016884469.1 |
Set Membership: |
HapMap |