Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAGGGCCGGATCCTTTTTAGCA[A/G]TACTTGTACTTTGCATTGGACTAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613888 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARGFXP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARGFXP2 - arginine-fifty homeobox pseudogene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371730 - uncharacterized LOC105371730 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOT1 - ras homolog family member T1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033566.2 | Intron | NP_001028738.1 | ||||
NM_001033567.2 | Intron | NP_001028739.2 | ||||
NM_001033568.2 | Intron | NP_001028740.1 | ||||
NM_001288754.1 | Intron | NP_001275683.1 | ||||
NM_001288755.1 | Intron | NP_001275684.1 | ||||
NM_001288758.1 | Intron | NP_001275687.1 | ||||
NM_018307.4 | Intron | NP_060777.3 | ||||
XM_011524969.2 | Intron | XP_011523271.1 | ||||
XM_011524971.2 | Intron | XP_011523273.2 | ||||
XM_011524973.2 | Intron | XP_011523275.2 | ||||
XM_011524975.2 | Intron | XP_011523277.1 | ||||
XM_011524976.2 | Intron | XP_011523278.2 | ||||
XM_017024819.1 | Intron | XP_016880308.1 | ||||
XM_017024820.1 | Intron | XP_016880309.1 | ||||
XM_017024821.1 | Intron | XP_016880310.1 | ||||
XM_017024822.1 | Intron | XP_016880311.1 | ||||
XM_017024823.1 | Intron | XP_016880312.1 | ||||
XM_017024824.1 | Intron | XP_016880313.1 | ||||
XM_017024825.1 | Intron | XP_016880314.1 |
Set Membership: |
HapMap |