Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609689 | ||||||||||||||||||||
Literature Links: |
CEP250 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP250 - centrosomal protein 250 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318219.1 | Intron | NP_001305148.1 | ||||
NM_007186.5 | Intron | NP_009117.2 | ||||
XM_005260262.4 | Intron | XP_005260319.1 | ||||
XM_005260263.4 | Intron | XP_005260320.1 | ||||
XM_005260264.4 | Intron | XP_005260321.1 | ||||
XM_006723690.3 | Intron | XP_006723753.1 | ||||
XM_006723691.1 | Intron | XP_006723754.1 | ||||
XM_006723692.3 | Intron | XP_006723755.1 | ||||
XM_006723693.3 | Intron | XP_006723756.1 | ||||
XM_006723694.3 | Intron | XP_006723757.1 | ||||
XM_011528517.2 | Intron | XP_011526819.1 | ||||
XM_011528518.2 | Intron | XP_011526820.1 | ||||
XM_011528519.2 | Intron | XP_011526821.1 | ||||
XM_017027617.1 | Intron | XP_016883106.1 | ||||
XM_017027618.1 | Intron | XP_016883107.1 | ||||
XM_017027619.1 | Intron | XP_016883108.1 |